Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGCAGGGTAGAATGAGAGGGGG[A/G]AAAACAAAAGACGTGAGAATCAAAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604859 | |||||||||||||||||||||||
Literature Links: |
LMCD1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LMCD1 - LIM and cysteine rich domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278233.1 | Intron | NP_001265162.1 | ||||
NM_001278234.1 | Intron | NP_001265163.1 | ||||
NM_001278235.1 | Intron | NP_001265164.1 | ||||
NM_014583.3 | Intron | NP_055398.1 | ||||
XM_011533649.1 | Intron | XP_011531951.1 |
LMCD1-AS1 - LMCD1 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |