Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCTGCACCTTCATTTCTTGAAG[A/G]CTGTTCATGATCTTCTTCTGGTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600004 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPHA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPHA5 - EPH receptor A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281765.2 | 3607 | Silent Mutation | AGC,AGT | S,S 1023 | NP_001268694.1 | |
NM_001281766.2 | 3607 | Silent Mutation | AGC,AGT | S,S 1001 | NP_001268695.1 | |
NM_001281767.2 | 3607 | Intron | NP_001268696.1 | |||
NM_001318761.1 | 3607 | Silent Mutation | AGC,AGT | S,S 954 | NP_001305690.1 | |
NM_004439.7 | 3607 | Silent Mutation | AGC,AGT | S,S 1022 | NP_004430.4 | |
NM_182472.4 | 3607 | Silent Mutation | AGC,AGT | S,S 1000 | NP_872272.2 | |
XM_005265653.3 | 3607 | Silent Mutation | AGC,AGT | S,S 859 | XP_005265710.1 | |
XM_011531735.2 | 3607 | Silent Mutation | AGC,AGT | S,S 911 | XP_011530037.1 | |
XM_017007878.1 | 3607 | Silent Mutation | AGC,AGT | S,S 889 | XP_016863367.1 | |
XM_017007879.1 | 3607 | Intron | XP_016863368.1 | |||
XM_017007880.1 | 3607 | Silent Mutation | AGC,AGT | S,S 837 | XP_016863369.1 | |
XM_017007881.1 | 3607 | Intron | XP_016863370.1 |