Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGCACATTTTAAGCAAAAGGAG[C/T]AGCAAGTGCAAAGGTGTGAAAACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608092 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PALLD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PALLD - palladin, cytoskeletal associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166108.1 | Intron | NP_001159580.1 | ||||
NM_001166109.1 | Intron | NP_001159581.1 | ||||
NM_001166110.1 | Intron | NP_001159582.1 | ||||
NM_016081.3 | Intron | NP_057165.3 | ||||
XM_005262861.4 | Intron | XP_005262918.1 | ||||
XM_005262866.2 | Intron | XP_005262923.1 | ||||
XM_011531768.2 | Intron | XP_011530070.1 | ||||
XM_011531769.2 | Intron | XP_011530071.1 | ||||
XM_011531770.2 | Intron | XP_011530072.1 | ||||
XM_011531771.2 | Intron | XP_011530073.1 | ||||
XM_011531772.2 | Intron | XP_011530074.1 | ||||
XM_011531773.1 | Intron | XP_011530075.1 | ||||
XM_011531774.1 | Intron | XP_011530076.1 | ||||
XM_011531775.1 | Intron | XP_011530077.1 | ||||
XM_017007910.1 | Intron | XP_016863399.1 | ||||
XM_017007911.1 | Intron | XP_016863400.1 |