Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCTAGGCAGTTTTGAACAAGGA[C/G]AGCCCCTCTGGTGGAATTCAAGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERGIC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERGIC1 - endoplasmic reticulum-golgi intermediate compartment 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031711.2 | Intron | NP_001026881.1 | ||||
XM_006714892.1 | Intron | XP_006714955.1 | ||||
XM_011534597.1 | Intron | XP_011532899.1 | ||||
XM_011534598.2 | Intron | XP_011532900.1 | ||||
XM_017009654.1 | Intron | XP_016865143.1 | ||||
XM_017009655.1 | Intron | XP_016865144.1 |