Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTTTTTTTATTAGAAAAGAAATT[A/T]AGTGTTGTCACTAGAGATTGAAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600443 | ||||||||||||||||||||
Literature Links: |
GLRX PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLRX - glutaredoxin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001118890.1 | 916 | UTR 3 | NP_001112362.1 | |||
NM_001243658.1 | 916 | UTR 3 | NP_001230587.1 | |||
NM_001243659.1 | 916 | UTR 3 | NP_001230588.1 | |||
NM_002064.2 | 916 | UTR 3 | NP_002055.1 |