Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTAATGAAGATGAGGAACGCTAA[A/G]TGTCTTTTTTAAATGGAGATGATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM124A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM124A - family with sequence similarity 124 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242312.1 | Intron | NP_001229241.1 | ||||
NM_145019.3 | Intron | NP_659456.3 | ||||
XM_011534978.2 | Intron | XP_011533280.1 | ||||
XM_017020419.1 | Intron | XP_016875908.1 | ||||
XM_017020420.1 | Intron | XP_016875909.1 |