Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACATTACATAGTATAAGGCTTAC[C/T]CAGACCAAGTTTGATCATCCTGTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 140571 | ||||||||||||||||||||
Literature Links: |
HSP90AA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSP90AA1 - heat shock protein 90 alpha family class A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017963.2 | 2279 | Missense Mutation | AGT,GGT | S,G 819 | NP_001017963.2 | |
NM_005348.3 | 2279 | Missense Mutation | AGT,GGT | S,G 697 | NP_005339.3 | |
XM_011536718.2 | 2279 | Missense Mutation | AGT,GGT | S,G 818 | XP_011535020.1 |