Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGGAGAAAAAGCTTCAGGGCTCC[A/G]GTGACCACATGAATGTCATCCCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610576 MIM: 165220 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP9 - Rho GTPase activating protein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080156.2 | 2311 | Silent Mutation | ACC,ACT | T,T 428 | NP_001073625.1 | |
NM_001080157.1 | 2311 | Silent Mutation | ACC,ACT | T,T 612 | NP_001073626.1 | |
NM_001319850.1 | 2311 | Silent Mutation | ACC,ACT | T,T 702 | NP_001306779.1 | |
NM_001319851.1 | 2311 | Silent Mutation | ACC,ACT | T,T 301 | NP_001306780.1 | |
NM_001319852.1 | 2311 | Silent Mutation | ACC,ACT | T,T 447 | NP_001306781.1 | |
NM_032496.3 | 2311 | Silent Mutation | ACC,ACT | T,T 612 | NP_115885.2 | |
XM_005269083.2 | 2311 | Silent Mutation | ACC,ACT | T,T 447 | XP_005269140.1 | |
XM_005269084.2 | 2311 | Silent Mutation | ACC,ACT | T,T 416 | XP_005269141.1 | |
XM_005269085.2 | 2311 | Silent Mutation | ACC,ACT | T,T 416 | XP_005269142.1 | |
XM_011538656.2 | 2311 | Silent Mutation | ACC,ACT | T,T 612 | XP_011536958.1 | |
XM_011538657.2 | 2311 | Silent Mutation | ACC,ACT | T,T 600 | XP_011536959.1 | |
XM_011538658.2 | 2311 | Silent Mutation | ACC,ACT | T,T 581 | XP_011536960.1 | |
XM_011538659.2 | 2311 | Silent Mutation | ACC,ACT | T,T 612 | XP_011536961.1 | |
XM_017019800.1 | 2311 | Silent Mutation | ACC,ACT | T,T 447 | XP_016875289.1 | |
XM_017019801.1 | 2311 | Silent Mutation | ACC,ACT | T,T 428 | XP_016875290.1 | |
XM_017019802.1 | 2311 | Silent Mutation | ACC,ACT | T,T 397 | XP_016875291.1 | |
XM_017019803.1 | 2311 | Silent Mutation | ACC,ACT | T,T 428 | XP_016875292.1 | |
XM_017019804.1 | 2311 | Silent Mutation | ACC,ACT | T,T 397 | XP_016875293.1 |
GLI1 - GLI family zinc finger 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |