Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTCAGGAAGCTCTGTTCCAGAA[C/T]AGAGGAATTTCTTAAAGGAAACGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601902 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CC2D1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CC2D1B - coiled-coil and C2 domain containing 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ORC1 - origin recognition complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190818.1 | 2437 | Missense Mutation | ATT,GTT | I,V 772 | NP_001177747.1 | |
NM_001190819.1 | 2437 | Missense Mutation | ATT,GTT | I,V 767 | NP_001177748.1 | |
NM_004153.3 | 2437 | Missense Mutation | ATT,GTT | I,V 772 | NP_004144.2 | |
XM_011541527.2 | 2437 | Missense Mutation | ATT,GTT | I,V 412 | XP_011539829.1 | |
XM_017001388.1 | 2437 | Missense Mutation | ATT,GTT | I,V 736 | XP_016856877.1 | |
XM_017001389.1 | 2437 | Missense Mutation | ATT,GTT | I,V 526 | XP_016856878.1 |
PLA2G12AP1 - phospholipase A2 group XIIA pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |