Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAAATTAACACCCACGAACACC[A/C]ATGGTATGTCACCTAAACCTAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603809 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH3D21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SH3D21 - SH3 domain containing 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162530.1 | Intron | NP_001156002.1 | ||||
NM_024676.4 | Intron | NP_078952.4 | ||||
XM_011542153.2 | Intron | XP_011540455.1 | ||||
XM_017002340.1 | Intron | XP_016857829.1 | ||||
XM_017002341.1 | Intron | XP_016857830.1 | ||||
XM_017002342.1 | Intron | XP_016857831.1 | ||||
XM_017002343.1 | Intron | XP_016857832.1 | ||||
XM_017002344.1 | Intron | XP_016857833.1 | ||||
XM_017002345.1 | Intron | XP_016857834.1 | ||||
XM_017002346.1 | Intron | XP_016857835.1 |
THRAP3 - thyroid hormone receptor associated protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |