Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCAGAAACCAGGGAACTTGGCTG[C/T]AAGTGTTACCTTTATTTATTTCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603430 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF202 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF202 - zinc finger protein 202 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301779.1 | 4196 | UTR 3 | NP_001288708.1 | |||
NM_001301780.1 | 4196 | UTR 3 | NP_001288709.1 | |||
NM_001301819.1 | 4196 | UTR 3 | NP_001288748.1 | |||
NM_003455.3 | 4196 | UTR 3 | NP_003446.2 | |||
XM_005271659.1 | 4196 | UTR 3 | XP_005271716.1 | |||
XM_005271660.1 | 4196 | UTR 3 | XP_005271717.1 | |||
XM_005271661.1 | 4196 | UTR 3 | XP_005271718.1 | |||
XM_005271664.1 | 4196 | UTR 3 | XP_005271721.1 | |||
XM_006718901.2 | 4196 | UTR 3 | XP_006718964.1 | |||
XM_011542972.1 | 4196 | UTR 3 | XP_011541274.1 | |||
XM_011542973.1 | 4196 | UTR 3 | XP_011541275.1 | |||
XM_011542975.1 | 4196 | UTR 3 | XP_011541277.1 | |||
XM_011542976.1 | 4196 | UTR 3 | XP_011541278.1 | |||
XM_017018268.1 | 4196 | UTR 3 | XP_016873757.1 |