Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGTCTCTTAGCTCTAGGAAAAAG[A/G]ATTTTGGAGGACAATATTGACCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614586 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZDHHC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZDHHC5 - zinc finger DHHC-type containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015457.2 | Intron | NP_056272.2 | ||||
XM_011544899.1 | Intron | XP_011543201.1 | ||||
XM_011544900.1 | Intron | XP_011543202.1 | ||||
XM_011544901.1 | Intron | XP_011543203.1 | ||||
XM_017017498.1 | Intron | XP_016872987.1 |