Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTAGATAATCACATATTAAAAAG[A/G]TAATGACTCATTATCAGTGAATTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606444 MIM: 612988 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CREBZF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CREBZF - CREB/ATF bZIP transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039618.2 | 3093 | UTR 3 | NP_001034707.1 | |||
XM_006718642.3 | 3093 | UTR 3 | XP_006718705.1 | |||
XM_011545195.2 | 3093 | UTR 3 | XP_011543497.1 | |||
XM_017018086.1 | 3093 | UTR 3 | XP_016873575.1 | |||
XM_017018087.1 | 3093 | UTR 3 | XP_016873576.1 | |||
XM_017018088.1 | 3093 | UTR 3 | XP_016873577.1 | |||
XM_017018089.1 | 3093 | UTR 3 | XP_016873578.1 | |||
XM_017018090.1 | 3093 | UTR 3 | XP_016873579.1 | |||
XM_017018091.1 | 3093 | UTR 3 | XP_016873580.1 | |||
XM_017018092.1 | 3093 | UTR 3 | XP_016873581.1 |
TMEM126A - transmembrane protein 126A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |