Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATTCCTCCGTCCTCCCACCACCT[A/C]ACTGTTCAGCTGTTGGTCAACACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612919 MIM: 611915 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LANCL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LANCL2 - LanC like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VOPP1 - vesicular, overexpressed in cancer, prosurvival protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284282.1 | Intron | NP_001271211.1 | ||||
NM_001284283.1 | Intron | NP_001271212.1 | ||||
NM_001284284.1 | Intron | NP_001271213.1 | ||||
NM_001321242.1 | Intron | NP_001308171.1 | ||||
NM_001321243.1 | Intron | NP_001308172.1 | ||||
NM_001321244.1 | Intron | NP_001308173.1 | ||||
NM_001321246.1 | Intron | NP_001308175.1 | ||||
NM_001321247.1 | Intron | NP_001308176.1 | ||||
NM_001321248.1 | Intron | NP_001308177.1 | ||||
NM_001321249.1 | Intron | NP_001308178.1 | ||||
NM_001321250.1 | Intron | NP_001308179.1 | ||||
NM_001321251.1 | Intron | NP_001308180.1 | ||||
NM_030796.4 | Intron | NP_110423.3 | ||||
XM_011515539.1 | Intron | XP_011513841.1 | ||||
XM_011515540.2 | Intron | XP_011513842.1 | ||||
XM_011515541.2 | Intron | XP_011513843.1 | ||||
XM_011515546.2 | Intron | XP_011513848.1 |