Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGCGCCTCCTGCTCACCTGGGTC[C/T]GCAGCTGCTGCTGTTCCCGCTCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600743 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01569 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC01569 - long intergenic non-protein coding RNA 1569 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105371063 - putative uncharacterized protein encoded by LINC00596 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TFAP4 - transcription factor AP-4 (activating enhancer binding protein 4) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003223.2 | 1049 | Missense Mutation | NP_003214.1 | |||
XM_011522633.2 | 1049 | Missense Mutation | XP_011520935.1 | |||
XM_011522635.2 | 1049 | Missense Mutation | XP_011520937.1 |