Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAAAAGGCAGGAGCCCACACAC[A/G]TCATCCATGCAGTGCACCTGCGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606007 MIM: 614403 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLR3K PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POLR3K - RNA polymerase III subunit K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHBDF1 - rhomboid 5 homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022450.3 | 2121 | Silent Mutation | GAC,GAT | D,D 637 | NP_071895.3 | |
XM_005255494.1 | 2121 | Silent Mutation | GAC,GAT | D,D 637 | XP_005255551.1 | |
XM_005255498.2 | 2121 | Silent Mutation | GAC,GAT | D,D 321 | XP_005255555.1 | |
XM_006720921.1 | 2121 | Silent Mutation | GAC,GAT | D,D 660 | XP_006720984.1 | |
XM_017023556.1 | 2121 | Silent Mutation | GAC,GAT | D,D 660 | XP_016879045.1 | |
XM_017023557.1 | 2121 | Silent Mutation | GAC,GAT | D,D 606 | XP_016879046.1 | |
XM_017023558.1 | 2121 | Intron | XP_016879047.1 |
SNRNP25 - small nuclear ribonucleoprotein U11/U12 subunit 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |