Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTCTGGCTGGGACTTTAGTTCCT[C/T]GTCCTCCAGCACTTGCACAGGGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
31 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604188 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SELENBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SELENBP1 - selenium binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258288.1 | 1401 | Missense Mutation | AAG,GAG | K,E 305 | NP_001245217.1 | |
NM_001258289.1 | 1401 | Missense Mutation | AAG,GAG | K,E 409 | NP_001245218.1 | |
NM_003944.3 | 1401 | Missense Mutation | AAG,GAG | K,E 367 | NP_003935.2 | |
XM_017002757.1 | 1401 | Intron | XP_016858246.1 |