Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGGCAGCAAATAAAAGGTGGAC[A/G]ACTCTGTCCCTCGATTTGCAGATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RUFY4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RUFY4 - RUN and FYVE domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198483.3 | Intron | NP_940885.2 | ||||
XM_011511014.2 | Intron | XP_011509316.1 | ||||
XM_017003895.1 | Intron | XP_016859384.1 | ||||
XM_017003896.1 | Intron | XP_016859385.1 | ||||
XM_017003897.1 | Intron | XP_016859386.1 |