Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAAATCATTTTCTTCTATGAAAA[C/T]TTTCTTCTAGAATAAATAAGATGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151443 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIFR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIFR - leukemia inhibitory factor receptor alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127671.1 | 7714 | UTR 3 | NP_001121143.1 | |||
NM_002310.5 | 7714 | UTR 3 | NP_002301.1 | |||
XM_011514040.2 | 7714 | UTR 3 | XP_011512342.1 | |||
XM_011514042.2 | 7714 | UTR 3 | XP_011512344.1 | |||
XM_017009462.1 | 7714 | UTR 3 | XP_016864951.1 | |||
XM_017009463.1 | 7714 | UTR 3 | XP_016864952.1 |