Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCGGCGAGCCAGGCGGCGGATAGC[A/G]GGCTTGGTGATTCCTTGGATATTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602792 MIM: 602805 MIM: 602811 MIM: 602823 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIST1H2AD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIST1H2AD - histone cluster 1, H2ad | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H2BE - histone cluster 1, H2be | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H3D - histone cluster 1, H3d | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H4D - histone cluster 1, H4d | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003539.3 | 99 | Silent Mutation | CCC,CCT | P,P 33 | NP_003530.1 |