Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCAGCTGGCATATTTCAGAGAT[G/T]GAGACAGCAGGTTATTTTCAGCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610082 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4639 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4639 - microRNA 4639 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYLIP - myosin regulatory light chain interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013262.3 | Intron | NP_037394.2 | ||||
XM_005249032.2 | Intron | XP_005249089.1 | ||||
XM_005249033.2 | Intron | XP_005249090.1 | ||||
XM_017010789.1 | Intron | XP_016866278.1 |