Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATTTCATTTTTCATCTGCACCC[C/T]CCAGGTTACATTTAAAACTGAAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176891 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRZ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTPRZ1 - protein tyrosine phosphatase, receptor type Z1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206838.1 | Intron | NP_001193767.1 | ||||
NM_001206839.1 | Intron | NP_001193768.1 | ||||
NM_002851.2 | Intron | NP_002842.2 | ||||
XM_005250519.1 | Intron | XP_005250576.1 | ||||
XM_017012477.1 | Intron | XP_016867966.1 |