Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACATTTAGGTAGGACCTACAGCT[C/T]GTCCGAGAGAGTAACTCATTTGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607312 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZC3HAV1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZC3HAV1 - zinc finger CCCH-type containing, antiviral 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020119.3 | 6580 | UTR 3 | NP_064504.2 | |||
NM_024625.3 | 6580 | Intron | NP_078901.3 | |||
XM_005250501.2 | 6580 | UTR 3 | XP_005250558.1 | |||
XM_011516412.2 | 6580 | Intron | XP_011514714.1 |
ZC3HAV1L - zinc finger CCCH-type containing, antiviral 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |