Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTCATTACTGTACTTGGAATTG[A/G]GACCAAATCAATATTTCTTAGTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM132B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM132B - transmembrane protein 132B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286219.1 | Intron | NP_001273148.1 | ||||
NM_052907.3 | Intron | NP_443139.2 | ||||
XM_011537852.2 | Intron | XP_011536154.1 | ||||
XM_011537854.2 | Intron | XP_011536156.1 | ||||
XM_017018767.1 | Intron | XP_016874256.1 |