Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACACAAGTACAAAGAATTACATAA[G/T]CTCCACATATTTATAATAGCTTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606572 MIM: 609399 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOX5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOX5 - NADPH oxidase, EF-hand calcium binding domain 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184779.1 | Intron | NP_001171708.1 | ||||
NM_001184780.1 | Intron | NP_001171709.1 | ||||
NM_024505.3 | Intron | NP_078781.3 |
SPESP1 - sperm equatorial segment protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145658.3 | Intron | NP_663633.1 |