Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGGCCACTTCTCTAGGCCAAGC[C/G]AAGCAGATTGAGAGAAGTTCCTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607512 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAMTS18 - ADAM metallopeptidase with thrombospondin type 1 motif 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001326358.1 | 5023 | UTR 3 | NP_001313287.1 | |||
NM_199355.3 | 5023 | UTR 3 | NP_955387.1 | |||
XM_011522923.1 | 5023 | UTR 3 | XP_011521225.1 | |||
XM_011522924.1 | 5023 | UTR 3 | XP_011521226.1 | |||
XM_017022988.1 | 5023 | UTR 3 | XP_016878477.1 | |||
XM_017022989.1 | 5023 | UTR 3 | XP_016878478.1 |