Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATTGTATTCACATGTGTTAACT[A/C]TGTAACTTCAAGTTCAGCATATAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADPRM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADPRM - ADP-ribose/CDP-alcohol diphosphatase, manganese dependent | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020233.4 | 1824 | UTR 3 | NP_064618.3 | |||
XM_005256738.2 | 1824 | Intron | XP_005256795.1 |
MAGOH2P - mago homolog 2, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM220 - transmembrane protein 220 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004313.1 | 1824 | Intron | NP_001004313.1 | |||
XM_005256634.4 | 1824 | Intron | XP_005256691.1 | |||
XM_011523835.2 | 1824 | Intron | XP_011522137.1 | |||
XM_011523836.2 | 1824 | Intron | XP_011522138.1 | |||
XM_017024616.1 | 1824 | UTR 3 | XP_016880105.1 | |||
XM_017024617.1 | 1824 | UTR 3 | XP_016880106.1 |