Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCTGTCGGTGGCCTGGGCTGGA[A/G]CAGTAGGAGTCCCCGGACCCAGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615695 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEXIM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HEXIM2 - hexamethylene bisacetamide inducible 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303436.1 | 1322 | Missense Mutation | AAC,AGC | N,S 90 | NP_001290365.1 | |
NM_001303437.1 | 1322 | Missense Mutation | AAC,AGC | N,S 68 | NP_001290366.1 | |
NM_001303438.1 | 1322 | Missense Mutation | AAC,AGC | N,S 68 | NP_001290367.1 | |
NM_001303439.1 | 1322 | Missense Mutation | AAC,AGC | N,S 68 | NP_001290368.1 | |
NM_001303440.1 | 1322 | Missense Mutation | AAC,AGC | N,S 68 | NP_001290369.1 | |
NM_001303441.1 | 1322 | Missense Mutation | AAC,AGC | N,S 68 | NP_001290370.1 | |
NM_001303442.1 | 1322 | Missense Mutation | AAC,AGC | N,S 68 | NP_001290371.1 | |
NM_001303443.1 | 1322 | Missense Mutation | AAC,AGC | N,S 68 | NP_001290372.1 | |
NM_001303444.1 | 1322 | Missense Mutation | AAC,AGC | N,S 68 | NP_001290373.1 | |
NM_144608.2 | 1322 | Missense Mutation | AAC,AGC | N,S 68 | NP_653209.1 | |
XM_006721687.3 | 1322 | Missense Mutation | AAC,AGC | N,S 91 | XP_006721750.1 | |
XM_011524306.2 | 1322 | Missense Mutation | AAC,AGC | N,S 68 | XP_011522608.1 | |
XM_011524307.2 | 1322 | Missense Mutation | AAC,AGC | N,S 228 | XP_011522609.2 | |
XM_011524308.2 | 1322 | Missense Mutation | AAC,AGC | N,S 229 | XP_011522610.2 | |
XM_017024167.1 | 1322 | Missense Mutation | AAC,AGC | N,S 68 | XP_016879656.1 | |
XM_017024168.1 | 1322 | Missense Mutation | AAC,AGC | N,S 68 | XP_016879657.1 | |
XM_017024169.1 | 1322 | Missense Mutation | AAC,AGC | N,S 68 | XP_016879658.1 | |
XM_017024170.1 | 1322 | Missense Mutation | AAC,AGC | N,S 68 | XP_016879659.1 |
LOC105371795 - uncharacterized LOC105371795 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |