Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTTGCAGGTGCCCTTCCGGGAC[A/G]CCTGGCCTGTAAGTGCTGCCCCCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602768 MIM: 607381 | ||||||||||||||||||||
Literature Links: |
DLL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DLL3 - delta like canonical Notch ligand 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016941.3 | 401 | Missense Mutation | ACC,GCC | T,A 115 | NP_058637.1 | |
NM_203486.2 | 401 | Missense Mutation | ACC,GCC | T,A 115 | NP_982353.1 |
TIMM50 - translocase of inner mitochondrial membrane 50 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |