Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTAAGAAAAAAAAAAAAAAAAA[A/G]AAAGGTCCCAAAGTGTAGGATCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610190 | ||||||||||||||||||||
Literature Links: |
CHST8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHST8 - carbohydrate sulfotransferase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127895.1 | Intron | NP_001121367.1 | ||||
NM_001127896.1 | Intron | NP_001121368.1 | ||||
NM_022467.3 | Intron | NP_071912.2 | ||||
XM_011527222.1 | Intron | XP_011525524.1 | ||||
XM_011527224.1 | Intron | XP_011525526.1 | ||||
XM_017027143.1 | Intron | XP_016882632.1 |