Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATATATCTACGTAAATAGAGAGA[A/G]ACTTCTAAATTAAAACATCCTGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601289 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
YWHAB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
YWHAB - tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003404.4 | Intron | NP_003395.1 | ||||
NM_139323.3 | Intron | NP_647539.1 | ||||
XM_017028039.1 | Intron | XP_016883528.1 | ||||
XM_017028040.1 | Intron | XP_016883529.1 |