Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCATATTGTGCAGTCCAAGGAGA[C/T]CCCTGGAGCCATAGCAATACTGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614272 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FASTKD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FASTKD5 - FAST kinase domains 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021826.4 | 2449 | Missense Mutation | NP_068598.1 |
UBOX5 - U-box domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267584.1 | 2449 | Intron | NP_001254513.1 | |||
NM_014948.3 | 2449 | Intron | NP_055763.1 | |||
NM_199415.2 | 2449 | Intron | NP_955447.1 |
UBOX5-AS1 - UBOX5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |