Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTAATAGAAATATCTCTTAAGGC[G/T]GGAATGGCCTCCTCTTGGTATTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605938 MIM: 602259 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105372795 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105372795 - uncharacterized LOC105372795 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGP - phosphatidylinositol glycan anchor biosynthesis class P | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320480.1 | 508 | Silent Mutation | CCA,CCC | P,P 109 | NP_001307409.1 | |
NM_016430.3 | 508 | Silent Mutation | CCA,CCC | P,P 83 | NP_057514.2 | |
NM_153681.2 | 508 | Silent Mutation | CCA,CCC | P,P 133 | NP_710148.1 | |
NM_153682.2 | 508 | Silent Mutation | CCA,CCC | P,P 109 | NP_710149.1 | |
XM_005260990.4 | 508 | Intron | XP_005261047.1 | |||
XM_011529596.2 | 508 | Silent Mutation | CCA,CCC | P,P 109 | XP_011527898.1 | |
XM_017028364.1 | 508 | Silent Mutation | CCA,CCC | P,P 109 | XP_016883853.1 | |
XM_017028365.1 | 508 | Silent Mutation | CCA,CCC | P,P 83 | XP_016883854.1 |
TTC3 - tetratricopeptide repeat domain 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |