Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCAGTACATTGCCTGCTGTGCTG[C/T]ACCACCCAGACCACCTCTTTGGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606590 MIM: 180073 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPLOC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NPLOC4 - NPL4 homolog, ubiquitin recognition factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDE6G - phosphodiesterase 6G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSPAN10 - tetraspanin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290212.1 | Intron | NP_001277141.1 | ||||
NM_031945.4 | Intron | NP_114151.3 |