Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTATGAAAAATCTCAGCTAACA[C/T]TCACTTTTTGGGAGTTAATCTTACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607033 MIM: 604709 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TFB1M PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TFB1M - transcription factor B1, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016020.3 | Intron | NP_057104.2 | ||||
XM_005267006.4 | Intron | XP_005267063.1 | ||||
XM_011535870.1 | Intron | XP_011534172.1 | ||||
XM_011535871.1 | Intron | XP_011534173.1 | ||||
XM_011535872.2 | Intron | XP_011534174.1 | ||||
XM_011535873.2 | Intron | XP_011534175.1 | ||||
XM_017010917.1 | Intron | XP_016866406.1 |
TIAM2 - T-cell lymphoma invasion and metastasis 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010927.2 | Intron | NP_001010927.1 | ||||
NM_012454.3 | Intron | NP_036586.2 |