Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTTAGAGTCATGGGAAAATGTG[A/G]TGAGGTTCCGAGTACTGCAGGTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616569 MIM: 147559 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSAD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSAD - cysteine sulfinic acid decarboxylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ITGB7 - integrin subunit beta 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF740 - zinc finger protein 740 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004304.3 | Intron | NP_001004304.1 | ||||
XM_006719344.2 | Intron | XP_006719407.1 | ||||
XM_011538194.2 | Intron | XP_011536496.1 | ||||
XM_017019181.1 | Intron | XP_016874670.1 |