Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGTAAAAGGAAAATGAGAATAA[C/T]GGGAGGGAAGAAGACAGGTTAAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609589 MIM: 604584 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTUS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTUS1 - microtubule associated tumor suppressor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001924.2 | 2159 | UTR 3 | NP_001001924.1 | |||
NM_001001925.2 | 2159 | UTR 3 | NP_001001925.1 | |||
NM_001001931.2 | 2159 | UTR 3 | NP_001001931.1 | |||
NM_001166393.1 | 2159 | UTR 3 | NP_001159865.1 | |||
NM_020749.4 | 2159 | UTR 3 | NP_065800.1 | |||
XM_005273577.3 | 2159 | UTR 3 | XP_005273634.1 | |||
XM_005273578.3 | 2159 | UTR 3 | XP_005273635.1 | |||
XM_005273579.3 | 2159 | UTR 3 | XP_005273636.1 | |||
XM_005273580.3 | 2159 | UTR 3 | XP_005273637.1 | |||
XM_005273581.3 | 2159 | UTR 3 | XP_005273638.1 | |||
XM_005273582.3 | 2159 | UTR 3 | XP_005273639.1 | |||
XM_005273583.4 | 2159 | Intron | XP_005273640.1 | |||
XM_011544595.2 | 2159 | UTR 3 | XP_011542897.1 | |||
XM_017013698.1 | 2159 | UTR 3 | XP_016869187.1 | |||
XM_017013699.1 | 2159 | UTR 3 | XP_016869188.1 | |||
XM_017013700.1 | 2159 | UTR 3 | XP_016869189.1 | |||
XM_017013701.1 | 2159 | UTR 3 | XP_016869190.1 | |||
XM_017013702.1 | 2159 | UTR 3 | XP_016869191.1 | |||
XM_017013703.1 | 2159 | Intron | XP_016869192.1 | |||
XM_017013704.1 | 2159 | UTR 3 | XP_016869193.1 |
PDGFRL - platelet derived growth factor receptor like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |