Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGACCTTGTAACCTTAGACAAC[C/T]TCTGAGCTTCTCTGAGCCTTGTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603886 MIM: 606494 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARTN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARTN - artemin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136215.1 | Intron | NP_001129687.1 | ||||
NM_057090.2 | Intron | NP_476431.2 | ||||
NM_057091.2 | Intron | NP_476432.2 |
ST3GAL3 - ST3 beta-galactoside alpha-2,3-sialyltransferase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |