Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGTCCGTTTTTAGGCTGTAATCG[G/T]CATTACTGTCAGCCAGTCAGCAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612141 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPB41L4A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPB41L4A - erythrocyte membrane protein band 4.1 like 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022140.3 | Intron | NP_071423.3 | ||||
XM_011543530.1 | Intron | XP_011541832.1 | ||||
XM_011543531.2 | Intron | XP_011541833.1 | ||||
XM_011543532.1 | Intron | XP_011541834.1 | ||||
XM_011543533.1 | Intron | XP_011541835.1 | ||||
XM_017009689.1 | Intron | XP_016865178.1 |
EPB41L4A-AS1 - EPB41L4A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA13 - small nucleolar RNA, H/ACA box 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |