Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAATGAAAACATTTTCAACTGCTT[A/G]TACAAAAACCCTGAGGCAACTTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OLAH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OLAH - oleoyl-ACP hydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039702.2 | 275 | Silent Mutation | TTA,TTG | L,L 19 | NP_001034791.1 | |
NM_018324.2 | 275 | Silent Mutation | TTA,TTG | L,L 19 | NP_060794.1 | |
XM_017016376.1 | 275 | Silent Mutation | TTA,TTG | L,L 19 | XP_016871865.1 |