Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATGGAGGAACTGAAGAAGGAAGT[A/G]GTCATGGTGAGGCCACCCAAAGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182340 MIM: 607321 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP1A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP1A2 - ATPase Na+/K+ transporting subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ATP1A4 - ATPase Na+/K+ transporting subunit alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001734.1 | 620 | Intron | NP_001001734.1 | |||
NM_144699.3 | 620 | Silent Mutation | GTA,GTG | V,V 47 | NP_653300.2 | |
XM_011509582.1 | 620 | Intron | XP_011507884.1 |