Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTGTCAGATCCTGTAAGATTTC[A/G]CTATTTTATCCAACATAAATTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611748 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTMR11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTMR11 - myotubularin related protein 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OTUD7B - OTU deubiquitinase 7B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020205.3 | 4750 | UTR 3 | NP_064590.2 | |||
XM_011509782.1 | 4750 | UTR 3 | XP_011508084.1 | |||
XM_011509783.2 | 4750 | UTR 3 | XP_011508085.1 | |||
XM_011509784.2 | 4750 | UTR 3 | XP_011508086.1 | |||
XM_011509785.2 | 4750 | UTR 3 | XP_011508087.1 | |||
XM_011509786.1 | 4750 | UTR 3 | XP_011508088.1 | |||
XM_011509787.1 | 4750 | UTR 3 | XP_011508089.1 | |||
XM_011509788.1 | 4750 | UTR 3 | XP_011508090.1 | |||
XM_017001850.1 | 4750 | UTR 3 | XP_016857339.1 | |||
XM_017001851.1 | 4750 | UTR 3 | XP_016857340.1 |