Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTGAACCATTAAGAAAAGATGGC[A/G]AAAGTCAACATAACTAGAGACCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDTC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDTC1 - WD and tetratricopeptide repeats 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276252.1 | 486 | Silent Mutation | GCA,GCG | A,A 2 | NP_001263181.1 | |
NM_015023.4 | 486 | Silent Mutation | GCA,GCG | A,A 2 | NP_055838.2 | |
XM_011541057.1 | 486 | Silent Mutation | GCA,GCG | A,A 2 | XP_011539359.1 | |
XM_011541058.2 | 486 | Silent Mutation | GCA,GCG | A,A 2 | XP_011539360.1 | |
XM_017000730.1 | 486 | Silent Mutation | GCA,GCG | A,A 2 | XP_016856219.1 |