Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATCTCCCTCCAGTACCTCAAGAA[C/T]GACACGTGGAGGCATACGTGTGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601405 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTRC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTRC - chymotrypsin C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007272.2 | 179 | Silent Mutation | AAC,AAT | N,N 52 | NP_009203.2 | |
XM_011540550.1 | 179 | Silent Mutation | AAC,AAT | N,N 52 | XP_011538852.1 |