Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTATGTCTTTACCATGTTTTCA[C/T]TCACTTAAAAAAAAGTGTTCTTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123811 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATF2 - activating transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256090.1 | 2316 | UTR 3 | NP_001243019.1 | |||
NM_001256091.1 | 2316 | UTR 3 | NP_001243020.1 | |||
NM_001256092.1 | 2316 | UTR 3 | NP_001243021.1 | |||
NM_001256093.1 | 2316 | UTR 3 | NP_001243022.1 | |||
NM_001256094.1 | 2316 | Intron | NP_001243023.1 | |||
NM_001880.3 | 2316 | UTR 3 | NP_001871.2 |