Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATAAAATTTAAACATAAATTTC[A/G]AGTGAACATTAATCTGATAGGGATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606575 MIM: 614423 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MPP4 - membrane palmitoylated protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033066.2 | Intron | NP_149055.1 | ||||
XM_017004620.1 | Intron | XP_016860109.1 |
TMEM237 - transmembrane protein 237 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |