Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACACAACACAAAAACTCCAGGTGT[G/T]CCCTGAGGCCTCTGCAGGGACCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611617 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFHD1 - EF-hand domain family member D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243252.1 | Intron | NP_001230181.1 | ||||
NM_001308395.1 | Intron | NP_001295324.1 | ||||
NM_025202.3 | Intron | NP_079478.1 | ||||
XM_011511924.2 | Intron | XP_011510226.1 |