Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606734 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OSBPL6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OSBPL6 - oxysterol binding protein like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201480.1 | Intron | NP_001188409.1 | ||||
NM_001201481.1 | Intron | NP_001188410.1 | ||||
NM_001201482.1 | Intron | NP_001188411.1 | ||||
NM_032523.3 | Intron | NP_115912.1 | ||||
NM_145739.2 | Intron | NP_665682.1 | ||||
XM_011510539.1 | Intron | XP_011508841.1 | ||||
XM_017003265.1 | Intron | XP_016858754.1 | ||||
XM_017003266.1 | Intron | XP_016858755.1 | ||||
XM_017003267.1 | Intron | XP_016858756.1 | ||||
XM_017003268.1 | Intron | XP_016858757.1 | ||||
XM_017003269.1 | Intron | XP_016858758.1 | ||||
XM_017003270.1 | Intron | XP_016858759.1 | ||||
XM_017003271.1 | Intron | XP_016858760.1 | ||||
XM_017003272.1 | Intron | XP_016858761.1 | ||||
XM_017003273.1 | Intron | XP_016858762.1 | ||||
XM_017003274.1 | Intron | XP_016858763.1 | ||||
XM_017003275.1 | Intron | XP_016858764.1 | ||||
XM_017003276.1 | Intron | XP_016858765.1 | ||||
XM_017003277.1 | Intron | XP_016858766.1 |