Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCCCTCCTTTGTTGGGGAGATC[C/T]ACAGAGAACATGTTAGTGGGAACTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615904 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PXDNL PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PXDNL - peroxidasin like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144651.4 | Intron | NP_653252.3 | ||||
XM_005251168.3 | Intron | XP_005251225.1 | ||||
XM_006716420.3 | Intron | XP_006716483.1 | ||||
XM_011517456.2 | Intron | XP_011515758.1 | ||||
XM_011517457.2 | Intron | XP_011515759.1 | ||||
XM_011517458.2 | Intron | XP_011515760.1 | ||||
XM_011517459.2 | Intron | XP_011515761.1 | ||||
XM_017013041.1 | Intron | XP_016868530.1 |
Set Membership: |
HapMap |