Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604665 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COPS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
COPS3 - COP9 signalosome subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199125.1 | Intron | NP_001186054.1 | ||||
NM_001316354.1 | Intron | NP_001303283.1 | ||||
NM_001316355.1 | Intron | NP_001303284.1 | ||||
NM_001316356.1 | Intron | NP_001303285.1 | ||||
NM_001316357.1 | Intron | NP_001303286.1 | ||||
NM_001316358.1 | Intron | NP_001303287.1 | ||||
NM_003653.3 | Intron | NP_003644.2 | ||||
XM_005256837.4 | Intron | XP_005256894.1 | ||||
XM_005256840.3 | Intron | XP_005256897.1 | ||||
XM_005256842.4 | Intron | XP_005256899.1 | ||||
XM_017025246.1 | Intron | XP_016880735.1 |
Set Membership: |
HapMap Validated |